Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

738816

Sigma-Aldrich

3-Methylglutaryl-L-carnitine-(methyl-13C,d3) hydrochloride

99 atom % 13C, 98 atom % D, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC12H21D3NO6Cl
Molecular Weight:
329.80
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

95% (CP)

form

gas

mass shift

M+4

storage temp.

−20°C

SMILES string

Cl.[2H][13C]([2H])([2H])[N+](C)(C)C[C@@H](CC([O-])=O)OC(=O)CC(C)CC(O)=O

InChI

1S/C13H23NO6.ClH/c1-9(5-11(15)16)6-13(19)20-10(7-12(17)18)8-14(2,3)4;/h9-10H,5-8H2,1-4H3,(H-,15,16,17,18);1H/t9?,10-;/m1./s1/i2+1D3;

InChI key

AUYDMXVJGDGHRP-NUGLJLLJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

2A - Compressed liquefied or pressurized gas

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service