Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

738220

Sigma-Aldrich

1,6,7,8-Tetrahydrocyclopenta[g]indole

97%

Synonym(s):

7,8-Dihydro-6H-cyclopenta[g]indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

80-85 °C

SMILES string

C1Cc2ccc3cc[nH]c3c2C1

InChI

1S/C11H11N/c1-2-8-4-5-9-6-7-12-11(9)10(8)3-1/h4-7,12H,1-3H2

InChI key

OGAMTYAYYRHIKY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service