Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

737887

Sigma-Aldrich

Benzylphosphonic acid

97%

Synonym(s):

Phenylmethylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9O3P
CAS Number:
Molecular Weight:
172.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

solid

mp

173-178 °C

SMILES string

OP(O)(=O)Cc1ccccc1

InChI

1S/C7H9O3P/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H2,8,9,10)

InChI key

OGBVRMYSNSKIEF-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nadine Borg et al.
Circulation, 136(3), 297-313 (2017-04-23)
T cells are required for proper healing after myocardial infarction. The mechanism of their beneficial action, however, is unknown. The proinflammatory danger signal ATP, released from damaged cells, is degraded by the ectonucleotidases CD39 and CD73 to the anti-inflammatory mediator
Organic-inorganic nanocomposites by placing conjugated polymers in intimate contact with quantum rods.
Lei Zhao et al.
Advanced materials (Deerfield Beach, Fla.), 23(25), 2844-2849 (2011-05-19)
D Elias Jesu Packiam et al.
Dalton transactions (Cambridge, England : 2003), 46(46), 16102-16112 (2017-11-11)
Twenty new molybdenum phosphonates and diphosphonates have been synthesized and structurally characterized by single crystal and powder X-ray diffraction, CHN analyses, spectroscopic and thermal studies. Four of them are molecular phenyl- and benzyl-phosphonates containing discrete [(MoO
Alejandra Duarte et al.
PloS one, 7(9), e45829-e45829 (2012-10-03)
Although the contribution of mitochondrial dynamics (a balance in fusion/fission events and changes in mitochondria subcellular distribution) to key biological process has been reported, the contribution of changes in mitochondrial fusion to achieve efficient steroid production has never been explored.
C Poderoso et al.
Endocrine research, 28(4), 413-417 (2003-01-18)
A key regulatory step in the steroidogenic hormones signaling pathway is the synthesis of steroidogenic acute regulatory protein (StAR). This protein facilitates the delivery of cholesterol to the inner mitochondrial membrane, the rate-limiting step in steroidogenesis. ACTH and LH pathway

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service