Skip to Content
MilliporeSigma
All Photos(1)

Documents

737852

Sigma-Aldrich

Nickel(II) hexafluorosilicate hexahydrate

99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NiSiF6 · 6H2O
CAS Number:
Molecular Weight:
308.86
EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.995% trace metals basis

SMILES string

O.O.O.O.O.O.[Ni++].F[Si--](F)(F)(F)(F)F

InChI

1S/F6Si.Ni.6H2O/c1-7(2,3,4,5)6;;;;;;;/h;;6*1H2/q-2;+2;;;;;;

InChI key

DRKXNSFDNAORRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shrivastava, K.N.
Chemical Physics Letters, 20, 106-106 (1973)
Wang, H.Q.; Kuang, X.-Y.; Li, H.-F.; Mao, A.-J.
J. Alloy Compounds, 468, 1-6 (2009)
Pryce, M.H.L.; Agnetta, G.; Carofano, T.; Palma-Vittorelli, M.B.; Palma, M.U.
Philosophical Magazine, 10, 477-477 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service