Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

736228

Supelco

Trimethyl phosphite solution

NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

concentration

1% in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in. , ultra-thin wall

NMR tube wall thickness

0.24 mm

format

single component solution

SMILES string

COP(OC)OC

InChI

1S/C3H9O3P/c1-4-7(5-2)6-3/h1-3H3

InChI key

CYTQBVOFDCPGCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

31P sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service