Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

735809

Sigma-Aldrich

[2-(Di-tert-butylphosphinomethyl)-6-(diethylaminomethyl)pyridine]carbonylchlorohydridoruthenium(II)

greener alternative

Synonym(s):

Milstein Catalyst Precursor, [2-(Di-tert-butylphosphinomethyl)-6-(diethylaminomethyl)pyridine]ruthenium(II) chlorocarbonyl hydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36ClN2OPRu
CAS Number:
Molecular Weight:
488.01
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

Cl[RuH].[C-]#[O+].CCN(CC)Cc1cccc(CP(C(C)(C)C)C(C)(C)C)n1

InChI

1S/C19H35N2P.CO.ClH.Ru.H/c1-9-21(10-2)14-16-12-11-13-17(20-16)15-22(18(3,4)5)19(6,7)8;1-2;;;/h11-13H,9-10,14-15H2,1-8H3;;1H;;/q;;;+1;/p-1

InChI key

RZCAJPSOQUIHSH-UHFFFAOYSA-M

General description

May contain up to 10% acetone and/or THF.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Precursor to active Ru catalyst for the dehydrogenative coupling of amines and alcohols to form amide bonds.
Precursor to active ruthenium catalyst for greener amide bond formation by dehydrogenative coupling of amines and alcohols.

Direct Synthesis of Amides from Alcohols and Amines with Liberation of H2

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service