Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

735566

Sigma-Aldrich

Ethyl (R)-1-Boc-4-oxopiperidine-2-carboxylate

97%

Synonym(s):

(R)-1-Boc-4-oxopiperidine-2-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$188.00

$188.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$188.00

About This Item

Empirical Formula (Hill Notation):
C13H21NO5
CAS Number:
Molecular Weight:
271.31
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$188.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

42-47 °C

functional group

ester
ketone

storage temp.

2-8°C

SMILES string

CCOC(=O)[C@H]1CC(=O)CCN1C(=O)OC(C)(C)C

InChI

1S/C13H21NO5/c1-5-18-11(16)10-8-9(15)6-7-14(10)12(17)19-13(2,3)4/h10H,5-8H2,1-4H3/t10-/m1/s1

InChI key

YAVQLRUBKDCOCI-SNVBAGLBSA-N

Application

Ethyl (R)-1-Boc-4-oxopiperidine-2-carboxylate can be used as a key intermediate in the preparation of 4-oxopipecolic acid enantiomer, a non-proteinogenic amino acid having antibiotic activity.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Practical synthesis of both enantiomers of protected 4-oxopipecolic acid
Machetti F, et al.
Tetrahedron, 57(23), 4995-4998 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service