Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

734608

Sigma-Aldrich

3,3′,5,5′-Tetrabromo-2,2′-bithiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H2Br4S2
CAS Number:
Molecular Weight:
481.85
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

138-143 °C

SMILES string

Brc1cc(Br)c(s1)-c2sc(Br)cc2Br

InChI

1S/C8H2Br4S2/c9-3-1-5(11)13-7(3)8-4(10)2-6(12)14-8/h1-2H

InChI key

MOMHMPZSZNZLAK-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service