Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

734454

Sigma-Aldrich

Thiophene-3-carbonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClOS
CAS Number:
Molecular Weight:
146.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

51-54 °C

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccsc1

InChI

1S/C5H3ClOS/c6-5(7)4-1-2-8-3-4/h1-3H

InChI key

QTWBEVAYYDZLQL-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

123.8 °F

Flash Point(C)

51 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service