Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

734047

Sigma-Aldrich

(S)-3-Aminotetrahydrofuran tosylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO4S
CAS Number:
Molecular Weight:
259.32
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]22/D −4.5°, c = 0.5 in methanol

mp

133-138 °C

storage temp.

2-8°C

SMILES string

N[C@H]1CCOC1.Cc2ccc(cc2)S(O)(=O)=O

InChI

1S/C7H8O3S.C4H9NO/c1-6-2-4-7(5-3-6)11(8,9)10;5-4-1-2-6-3-4/h2-5H,1H3,(H,8,9,10);4H,1-3,5H2/t;4-/m.0/s1

InChI key

BZXPLADBSZWDIH-VWMHFEHESA-N

Application

(S)-3-Aminotetrahydrofuran tosylate can be used as a reagent in the synthesis of oxadiazolyl-quinolinyl-N-(pyranyl)piperidinamines as potential κ-opioid receptor (KOR) antagonists. It may also be used in the preparation of N-(4-fluoro-3-methyl-phenyl)-3-[[(3S)-tetrahydrofuran-3-yl]-sulfamoyl]benzamide derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis of a novel and selective kappa opioid receptor (KOR) antagonist (BTRX-335140)
Guerrero M, et al.
Journal of Medicinal Chemistry, 61(14), 6247-6260 (2019)
Synthesis and evaluation of N-phenyl-3-sulfamoyl-benzamide derivatives as capsid assembly modulators inhibiting hepatitis B virus (HBV)
Vandyck K, et al.
Journal of medicinal chemistry, 61(14), 6247-6260 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service