Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733830

Sigma-Aldrich

Pyruvic-13C3 acid

99 atom % 13C, 95% (CP)

Synonym(s):

α-Ketopropionic acid-13C3, 2-Oxopropionic acid-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CO13COOH
CAS Number:
Molecular Weight:
91.04
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Assay

95% (CP)

form

liquid

bp

165 °C (lit.)

mp

11-12 °C (lit.)

density

1.309 g/mL at 25 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

[13CH3][13C](=O)[13C](O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)/i1+1,2+1,3+1

InChI key

LCTONWCANYUPML-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service