Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733660

Sigma-Aldrich

2-[(1′,1′,1′-Trifluoro-2′-(trifluoromethyl)-2′-hydroxy)propyl]-3-norbornyl methacrylate

>97%

Synonym(s):

3-[3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propyl]bicyclo[2.2.1]hept-2-yl methacrylate, HFA monomer

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$798.00

$798.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$798.00

About This Item

Empirical Formula (Hill Notation):
C15H18F6O3
CAS Number:
Molecular Weight:
360.29
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$798.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>97%

form

solid

mp

45-50 °C

SMILES string

CC(=C)C(=O)OC1C2CCC(C2)C1CC(O)(C(F)(F)F)C(F)(F)F

InChI

1S/C15H18F6O3/c1-7(2)12(22)24-11-9-4-3-8(5-9)10(11)6-13(23,14(16,17)18)15(19,20)21/h8-11,23H,1,3-6H2,2H3

InChI key

VRGGMPGFYAQUIC-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

To keep pace with Moore′s Law, there is a continuing need in the semiconductor industry to achieve higher circuit density in microelectronic devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service