Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733407

Sigma-Aldrich

Poly(ethylene glycol) methyl ether azide

PEG average Mn 1,000

Synonym(s):

Methoxypolyethylene glycol azide, α-Methoxy-ω-azido-PEG, O-(2-Azidoethyl)-O′-methylpolyethylene glycol, PEG-Azide, mPEG Azide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N3CH2CH2(OCH2CH2)nOCH3
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PEG average Mn 1,000

reaction suitability

reagent type: chemical modification reagent
reaction type: click chemistry

transition temp

Tm 37-41 °C

Mw/Mn

<1.1

Ω-end

azide

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service