Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733083

Sigma-Aldrich

(S,S)-1,3-Bis(4-bromobenzyl)-2-chlorooctahydro-2-(2E)-crotyl-1H-1,3,2-benzodiazasilole

Synonym(s):

(3aS,7aS)-1,3-Bis[(4-bromophenyl)methyl]-2-(2E)-2-buten-1-yl-2-chlorooctahydro-1H-1,3,2-benzodiazasilole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29Br2ClN2Si
CAS Number:
Molecular Weight:
568.85
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

mp

92-96 °C

SMILES string

C\C=C\C[Si]1(Cl)N(Cc2ccc(Br)cc2)[C@H]3CCCC[C@@H]3N1Cc4ccc(Br)cc4

InChI

1S/C24H29Br2ClN2Si/c1-2-3-16-30(27)28(17-19-8-12-21(25)13-9-19)23-6-4-5-7-24(23)29(30)18-20-10-14-22(26)15-11-20/h2-3,8-15,23-24H,4-7,16-18H2,1H3/b3-2+/t23-,24-/m0/s1

InChI key

ZRWSDIWSVXXBNG-KOAGWOQNSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service