Skip to Content
MilliporeSigma
All Photos(1)

Documents

732958

Sigma-Aldrich

7-Fluorochromone-2-carboxylic acid

Synonym(s):

7-Fluoro-4-oxo-1(4H)-benzopyran-2-carboxylic acid, 7-Fluoro-4-oxo-4H-chromene-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5FO4
CAS Number:
Molecular Weight:
208.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

239-245 °C

SMILES string

OC(=O)C1=CC(=O)c2ccc(F)cc2O1

InChI

1S/C10H5FO4/c11-5-1-2-6-7(12)4-9(10(13)14)15-8(6)3-5/h1-4H,(H,13,14)

InChI key

NOSMZQCJSBUIMM-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of fluorocarboxychromone aminopiperidine-based melanin-concentrating hormone receptor 1 antagonists

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service