Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732877

Sigma-Aldrich

6-Bromo-3-iodochromone

Synonym(s):

6-Bromo-3-iodo-4H-benzopyran-4-one, 6-Bromo-3-iodo-4H-chromen-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4BrIO2
CAS Number:
Molecular Weight:
350.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

143-149 °C

SMILES string

Brc1ccc2OC=C(I)C(=O)c2c1

InChI

1S/C9H4BrIO2/c10-5-1-2-8-6(3-5)9(12)7(11)4-13-8/h1-4H

InChI key

JAKJUHSVDDDAAC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service