Skip to Content
MilliporeSigma
All Photos(1)

Documents

732222

Sigma-Aldrich

4-Bromothiophene-3-carboxamide

97%

Synonym(s):

4-Bromothiophene-3-carboxylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BrNOS
CAS Number:
Molecular Weight:
206.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

150-155 °C

SMILES string

NC(=O)c1cscc1Br

InChI

1S/C5H4BrNOS/c6-4-2-9-1-3(4)5(7)8/h1-2H,(H2,7,8)

InChI key

BMZBDWPOBVGKSU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandon C Fillmore et al.
ACS omega, 5(36), 22914-22925 (2020-09-22)
A pathway to a range of diverse heterocycles was developed using a nucleophilic cyclization strategy. Lactams and ene-imines are accessed in a few steps from a common precursor, and these moieties are further elaborated to directly provide pyrroles or pyridines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service