Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731951

Sigma-Aldrich

3,6-Dibromo-9-ethylcarbazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11Br2N
CAS Number:
Molecular Weight:
353.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

137-142 °C

SMILES string

CCn1c2ccc(Br)cc2c3cc(Br)ccc13

InChI

1S/C14H11Br2N/c1-2-17-13-5-3-9(15)7-11(13)12-8-10(16)4-6-14(12)17/h3-8H,2H2,1H3

InChI key

GZBJRMVGNVDUCO-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service