Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731684

Sigma-Aldrich

2-(Tributylstannyl)-3-methoxypyrazine

97%

Synonym(s):

2-Methoxy-3-(tributylstannyl)pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H32N2OSn
CAS Number:
Molecular Weight:
399.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.515

density

1.167 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1nccnc1OC

InChI

1S/C5H5N2O.3C4H9.Sn/c1-8-5-4-6-2-3-7-5;3*1-3-4-2;/h2-3H,1H3;3*1,3-4H2,2H3;

InChI key

MEKPDHIMYRRQFX-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Rüdiger et al.
Cell reports, 32(6), 108015-108015 (2020-08-14)
Endothelial tube formation on a reconstituted basement membrane (Matrigel) is a well-established in vitro model for studying the processes of angiogenesis and vasculogenesis. However, to date, the organizing principles that underlie the morphogenesis of this network and that shape the initial
Ying Shen et al.
Cancer cell, 37(6), 800-817 (2020-06-10)
Tumors are influenced by the mechanical properties of their microenvironment. Using patient samples and atomic force microscopy, we found that tissue stiffness is higher in liver metastases than in primary colorectal tumors. Highly activated metastasis-associated fibroblasts increase tissue stiffness, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service