Skip to Content
MilliporeSigma
All Photos(1)

Documents

731617

Sigma-Aldrich

Potassium 2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3BF7K
CAS Number:
Molecular Weight:
270.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

90%

form

solid

mp

278-291 °C

storage temp.

2-8°C

SMILES string

[K+].Fc1cc(ccc1[B-](F)(F)F)C(F)(F)F

InChI

1S/C7H3BF7.K/c9-6-3-4(7(10,11)12)1-2-5(6)8(13,14)15;/h1-3H;/q-1;+1

InChI key

NKFIFIUHQRJHJP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service