Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

730556

Sigma-Aldrich

N-Iodomorpholine hydriodide

97%

Synonym(s):

N-Iodomorpholinium iodide, Morpholine-iodide adduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9I2NO
CAS Number:
Molecular Weight:
340.93
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

92-96 °C

storage temp.

2-8°C

SMILES string

I.IN1CCOCC1

InChI

1S/C4H8INO.HI/c5-6-1-3-7-4-2-6;/h1-4H2;1H

InChI key

NZOIUCVTBFWRCS-UHFFFAOYSA-N

Application

N-Iodomorpholine hydriodide is an iodinating reagent that can be used in:
  • The iodination of (4-ethynylphenyl)diphenylphosphine oxide to yield (4-(iodoethynyl)phenyl)diphenylphosphine oxide, which is further used to prepare phosphine oxide-iodotriazole hybrid molecules.
  • The synthesis of silver bis(pyrazolyl)-methane complex based supramolecular architectures.
  • The synthesis of iodo-1,4-naphthoquinones, which are used to prepare biologically significant 3-aryl-1,4-naphthoquinones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and biological activities of 3-aryl-1, 4-naphthoquinones-green palladium-catalysed Suzuki cross coupling
Louvis AR, et al.
New. J. Chem., 40(9), 7643-7656 (2016)
Cooperative binding in a phosphine oxide-based halogen bonded dimer drives supramolecular oligomerization
Maugeri L, et al.
The Journal of Organic Chemistry, 82(4), 1986-1995 (2017)
Supramolecular Assemblies in Silver Complexes: Phase Transitions and the Role of the Halogen Bond
Bonfant G, et al.
Inorganic Chemistry, 59(6), 4140-4149 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service