Skip to Content
MilliporeSigma
All Photos(1)

Documents

730416

Sigma-Aldrich

S-(α-Fluorobenzyl)-S-phenyl-N-(p-tolylsulfonyl)sulfoximine

97%

Synonym(s):

N-[(fluorophenylmethyl)oxidophenyl-λ4-sulfanylidene]-4-methyl-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18FNO3S2
CAS Number:
Molecular Weight:
403.49
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

148-154 °C

SMILES string

Cc1ccc(cc1)S(=O)(=O)N=S(=O)(C(F)c2ccccc2)c3ccccc3

InChI

1S/C20H18FNO3S2/c1-16-12-14-19(15-13-16)27(24,25)22-26(23,18-10-6-3-7-11-18)20(21)17-8-4-2-5-9-17/h2-15,20H,1H3

InChI key

ZAGQEXRRSCLLFW-UHFFFAOYSA-N

Application

S-(α-Fluorobenzyl)-S-phenyl-N-(p-tolylsulfonyl)sulfoximine can be used as a reagent for the synthesis of monofluorinated epoxides byO-cyclization reaction with ketones in the presence of a strong base.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Synthesis and Ring-Opening Reactions of Monofluorinated Epoxides Derived from α-Fluorosulfoximines
Zhang W and Hu J
advanced synthesis and catalysis, 352(16), 2799-2804 (2010)
Efficient Synthesis and Ring-Opening Reactions of Monofluorinated Epoxides Derived from α-Fluorosulfoximines
Zhang W and Hu J
Advanced Synthesis & Catalysis, 352(16), 2799-2804 (2010)
Efficient Synthesis and Ring-Opening Reactions of Monofluorinated Epoxides Derived from a-Fluorosulfoximines
Zhang, W.; Hu, J.
Advanced Synthesis & Catalysis, 352, 2799-2804 (2010)

Related Content

The major research interests of Prof. Jinbo Hu's lab include the development of new fluorination reagents and reactions, especially the difluoromethylation, difluoromethylenation, and monofluoromethylation methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service