Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

729280

Sigma-Aldrich

9,9-Dimethylfluorene-2-boronic acid pinacol ester

95%

Synonym(s):

2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, FL-BE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25BO2
CAS Number:
Molecular Weight:
320.23
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

130-135 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc3-c4ccccc4C(C)(C)c3c2

InChI

1S/C21H25BO2/c1-19(2)17-10-8-7-9-15(17)16-12-11-14(13-18(16)19)22-23-20(3,4)21(5,6)24-22/h7-13H,1-6H3

InChI key

DAZFRJAIIUPRQZ-UHFFFAOYSA-N

Application

9,9-Dimethylfluorene-2-boronic acid pinacol ester can be used in the synthesis of 1,2-diphenylindolizine derivatives for potential usage in organic light emitting diodes (OLEDs). It can also be used in the synthesis of pyrene-fluorene based chromophores for applications in photocatalysis and organic electronic devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and photophysical properties of 1, 2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device
Song YR, et al.
Luminescence, 31(2), 364-371 (2016)
Synthesis and spectral properties of a hexameric pyrene-fluorene chromophore based on cyclotriphosphazene
Alidaugi HA, et al.
Polyhedron, 81(2), 436-441 (2014)
Synthesis and spectral properties of fluorene substituted cyclic and polymeric phosphazenes
Alidaugi HA, et al.
Inorgorganica Chimica Acta, 457(2), 95-102 (2017)

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service