Skip to Content
MilliporeSigma
All Photos(1)

Documents

729213

Sigma-Aldrich

2-Vinylpyridine-d7

(stabilized with 4-tert-butylcatechol), 97 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D7N
CAS Number:
Molecular Weight:
112.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

97% (CP)

form

liquid

bp

79-82 °C/29 mmHg (lit.)

density

1.038 at 25 °C

mass shift

M+7

storage temp.

−20°C

SMILES string

[2H]\C([2H])=C(/[2H])c1nc([2H])c([2H])c([2H])c1[2H]

InChI

1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2/i1D2,2D,3D,4D,5D,6D

InChI key

KGIGUEBEKRSTEW-QMUWOJKRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F

Flash Point(C)

48 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service