Skip to Content
MilliporeSigma
All Photos(1)

Documents

728268

Sigma-Aldrich

(Iodoethynyl)benzene

≥97.0% (HPLC)

Synonym(s):

1-Iodo-2-phenylacetylene, 1-Iodo-2-phenylethyne, Phenylethynyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5I
CAS Number:
Molecular Weight:
228.03
Beilstein:
1905735
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

liquid

storage temp.

−20°C

SMILES string

IC#Cc1ccccc1

InChI

1S/C8H5I/c9-7-6-8-4-2-1-3-5-8/h1-5H

InChI key

VOHWEKIUKSUIDT-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stepan S Tatevosyan et al.
The Journal of organic chemistry, 85(12), 7863-7876 (2020-05-23)
An efficient direct approach to triazole-fused sultams has been developed. The key step of the proposed strategy is base-mediated cyclization of sulfonamide-tethered 5-iodo-1,2,3-triazoles which are readily available via an improved protocol for Cu-catalyzed 1,3-dipolar cycloaddition. The annulation of the sultam

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service