Skip to Content
MilliporeSigma
All Photos(1)

Documents

727539

Sigma-Aldrich

4-Amino-2-bromophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrNO
CAS Number:
Molecular Weight:
188.02
Beilstein:
2802564
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥96.5% (GCF)
97%

shelf life

expiry date on the label

SMILES string

Nc1ccc(O)c(Br)c1

InChI

1S/C6H6BrNO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H,8H2

InChI key

CBQJZWGBFZAUEV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew B Prendergast et al.
Physical chemistry chemical physics : PCCP, 18(6), 4320-4332 (2015-10-29)
Gas-phase product detection studies of o-hydroxyphenyl radical and O2 are reported at 373, 500, and 600 K, at 4 Torr (533.3 Pa), using VUV time-resolved synchrotron photoionisation mass spectrometry. The dominant products are assigned as o-benzoquinone (C6H4O2, m/z 108) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service