Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727407

Sigma-Aldrich

HDMC

≥95%

Synonym(s):

6-HDMCB, N-[(5-Chloro-3-oxido-1H-benzotriazol-1-yl)-4-morpholinylmethylene]-N-methylmethanaminium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17ClF6N5O2P
CAS Number:
Molecular Weight:
455.72
Beilstein:
11222273
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

reaction suitability

reaction type: Coupling Reactions

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.C\[N+](C)=C(\N1CCOCC1)n2n[n+]([O-])c3cc(Cl)ccc23

InChI

1S/C13H17ClN5O2.F6P/c1-16(2)13(17-5-7-21-8-6-17)18-11-4-3-10(14)9-12(11)19(20)15-18;1-7(2,3,4,5)6/h3-4,9H,5-8H2,1-2H3;/q+1;-1

InChI key

MGRTWUAUKJMOER-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service