Skip to Content
MilliporeSigma
All Photos(1)

Documents

727199

Sigma-Aldrich

(S)-3-Methoxy-α-methylbenzylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-1-(3-Methoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21
Beilstein:
4798959
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

SMILES string

COc1cccc(c1)[C@H](C)N

InChI

1S/C9H13NO/c1-7(10)8-4-3-5-9(6-8)11-2/h3-7H,10H2,1-2H3/t7-/m0/s1

InChI key

CJWGCBRQAHCVHW-ZETCQYMHSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

238.1 °F

Flash Point(C)

114.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service