Skip to Content
MilliporeSigma
All Photos(1)

Documents

726540

Sigma-Aldrich

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-(+)-1-AminoTetralin, (S)-(+)-1-Aminotetraline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H11)NH2
CAS Number:
Molecular Weight:
147.22
Beilstein:
2360279
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

produced by BASF

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.565

bp

250 °C

density

1.010 g/mL at 25 °C

SMILES string

N[C@H]1CCCc2ccccc12

InChI

1S/C10H13N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10H,3,5,7,11H2/t10-/m0/s1

InChI key

JRZGPXSSNPTNMA-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks, or chiral auxiliaries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service