Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725609

Sigma-Aldrich

4-[N-(Methylaminoethyl)aminomethyl]styrene

>90%

Synonym(s):

N-(4-Vinylbenzyl)-N′-methylethane-1,2-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2
CAS Number:
Molecular Weight:
190.28
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

>90%

form

liquid

refractive index

n20/D 1.553

density

1.009 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CNCCNCc1ccc(C=C)cc1

InChI

1S/C12H18N2/c1-3-11-4-6-12(7-5-11)10-14-9-8-13-2/h3-7,13-14H,1,8-10H2,2H3

InChI key

CBCWMRSMUMTGAX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service