Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725390

Sigma-Aldrich

1,1′-Bis[bis(diethylamino)phosphino]ferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H48FeN4P2
CAS Number:
Molecular Weight:
534.48
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: iron
reagent type: catalyst

refractive index

n20/D 1.565

density

1.101 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Fe].CCN(CC)P([C]1[CH][CH][CH][CH]1)N(CC)CC.CCN(CC)P([C]2[CH][CH][CH][CH]2)N(CC)CC

InChI

1S/2C13H24N2P.Fe/c2*1-5-14(6-2)16(15(7-3)8-4)13-11-9-10-12-13;/h2*9-12H,5-8H2,1-4H3;

InChI key

NXWJNPUBBUBMBN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.9 °F

Flash Point(C)

68.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service