Skip to Content
MilliporeSigma
All Photos(1)

Documents

724742

Sigma-Aldrich

1-Naphthalenethiol

99%

Synonym(s):

α-Naphthalenethiol, 1-Mercaptonaphthalene, 1-Naphthylthiol, Naphthalene-1-thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8S
CAS Number:
Molecular Weight:
160.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

density

1.158 g/mL at 25 °C

SMILES string

Sc1cccc2ccccc12

InChI

1S/C10H8S/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H

InChI key

SEXOVMIIVBKGGM-UHFFFAOYSA-N

General description

1-Naphthalenethiol (1-NAT) is a standard analyte that can be diffused in the polymeric network for the detection of optical properties.

Application

1-NAT can be used as a model analyte to characterize the performance of the surface enhanced raman scattering (SERS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lance E Talbert et al.
Journal of the American Society for Mass Spectrometry, 29(9), 1760-1767 (2018-04-07)
Ultraviolet photodissociation or UVPD is an increasingly popular option for tandem-mass spectrometry experiments. UVPD can be carried out at many wavelengths, and it is important to understand how the results will be impacted by this choice. Here, we explore the
Selective isolation and noninvasive analysis of circulating cancer stem cells through Raman imaging.
Cho H, et al.
Biosensors And Bioelectronics, 102(1), 372-382 (2018)
Zeptomol detection through controlled ultrasensitive surface-enhanced Raman scattering
Rodriguez-Lorenzo L, et al.
Journal of the American Chemical Society, 131(13), 4616-4618 (2009)
Surface enhanced Raman scattering using star-shaped gold colloidal nanoparticles
Rodriguez-Lorenzo L, et al.
The Journal of Physical Chemistry C, 114(16), 7336-7340 (2009)
Au@ pNIPAM colloids as molecular traps for surface-enhanced, spectroscopic, ultra-sensitive analysis.
Alvarez?Puebla RA, et al.
Angewandte Chemie (International Edition in English), 121(1), 144-149 (2009)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service