Skip to Content
MilliporeSigma
All Photos(1)

Documents

724475

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-methacrylic acid-co-octadecyl acrylate)

5 mol % in methacrylic acid, 1 mol % in octadecyl acrylate, Mn 30,000-60,000

Synonym(s):

Poly(NIPAM-co-MAA-co-ODA), Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)m (C4H6O2)n (C21H40O2)p
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

Mn 30,000-60,000

concentration

1 mol % in octadecyl acrylate
5 mol % in methacrylic acid

mp

>300 °C

General description

Intelligent Swelling/Collapsing copolymer that can be used as a temperature- and pH-sensitive material.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Quijada-Garrido; I.; Prior-Cabanillas; A.; Garrido; L.; Barrales-Rienda; J. M.
Macromolecules, 38, 7434-7434 (2005)
Salgado-Rodriguez; R.; Licea-Claverie; A.; Arndt; K. F.
Eur. Polymer J., 40, 1931-1931 (2004)

Articles

Poly(N-isopropylacrylamide), or PNIPAM, is a stimuli-responsive polymer that responds to changes in pH and temperature and has a LCST around 32 C.

Tissue engineering has become a key therapeutic tool in the treatment of damaged or diseased organs and tissues, such as blood vessels and urinary bladders.

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service