Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

724394

Sigma-Aldrich

1-Butyl-3-methylimidazolium methanesulfonate

Synonym(s):

1-Butyl-3-methyl-1H-imidazolium methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O3S
CAS Number:
Molecular Weight:
234.32
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

99.9% trace metals basis

mp

75-80 °C (lit.)

SMILES string

CS([O-])(=O)=O.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.CH4O3S/c1-3-4-5-10-7-6-9(2)8-10;1-5(2,3)4/h6-8H,3-5H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

PUHVBRXUKOGSBC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Capacitor technology, sensors, optical devices, magnetic fluids

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

246.2 °F

Flash Point(C)

119 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliveira, Flavia C. C.; Rossi, Liane M.; Jardim, Renato F.; Rubim, Joel C.
The Journal of Physical Chemistry C, 113, 8566-8566 (2009)
Marijana Blesic et al.
Physical chemistry chemical physics : PCCP, 11(39), 8939-8948 (2010-05-08)
A set of 1-alkyl-3-methylimidazolium alkanesulfonate ionic liquids, [C(n)mim][C(k)SO(3)], formed by the variation of the alkyl chain lengths both in the cation and the anion (n = 1-6, 8, or 10; k = 1-4, or 6), was synthesised, with sixteen of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service