Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723983

Sigma-Aldrich

Potassium indole-6-trifluoroborate

95%

Synonym(s):

Potassium 6-indolyltrifluoroborate, Potassium indolyl-6-trifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BF3KN
Molecular Weight:
223.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

297-302 °C

SMILES string

[K+].F[B-](F)(F)c1ccc2cc[nH]c2c1

InChI

1S/C8H6BF3N.K/c10-9(11,12)7-2-1-6-3-4-13-8(6)5-7;/h1-5,13H;/q-1;+1

InChI key

QRRROZIXUXOCTN-UHFFFAOYSA-N

Application

Potassium indole-6-trifluoroborate can be used as a reactant to prepare:
  • Indole derivatives by reacting with various aryl halides via palladium-catalyzed Suzuki-Miyaura cross-coupling reaction.[1]
  • (1H-indol-6-yl)boronic acid by hydrolysis using silica gel and water.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient hydrolysis of organotrifluoroborates via silica gel and water
Molander GA, et al.
The Journal of Organic Chemistry, 74(19), 7364-7369 (2009)
Scope of the Suzuki- Miyaura cross-coupling reactions of potassium heteroaryltrifluoroborates
Molander GA, et al.
The Journal of Organic Chemistry, 74(3), 973-980 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service