Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723940

Sigma-Aldrich

3-Bromo-1-(2-tetrahydropyranyl)-1H-pyrazole

95%

Synonym(s):

3-Bromo-1-(2-tetrahydropyranyl)pyrazole, 3-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11BrN2O
CAS Number:
Molecular Weight:
231.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.538

density

1.460 g/mL at 25 °C

SMILES string

Brc1ccn(n1)C2CCCCO2

InChI

1S/C8H11BrN2O/c9-7-4-5-11(10-7)8-3-1-2-6-12-8/h4-5,8H,1-3,6H2

InChI key

DFCAINPSKAFWCM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service