Skip to Content
MilliporeSigma
All Photos(1)

Documents

723509

Sigma-Aldrich

Cesium dodecafluoro-closo-dodecaborate

97%

Synonym(s):

Cesium dodecafluorododecaborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B12Cs2F12
Molecular Weight:
623.52
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: boron
reagent type: catalyst

mp

>600 °C (dec.)

SMILES string

[Cs+].[Cs+].FB1C[B-]2(F)B(F)[B-](F)(B(F)B(F)B(F)B(F)B(F)B1F)B2F

InChI

1S/CH2B11F11.2Cs/c13-2-1-11(22)9(20)12(23,10(11)21)8(19)7(18)6(17)5(16)4(15)3(2)14;;/h1H2;;/q-2;2*+1

InChI key

VORBFFRCZXMLGL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service