Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723223

Sigma-Aldrich

Dodecyl 2-bromoisobutyrate

Synonym(s):

Dodecyl initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H31BrO2
CAS Number:
Molecular Weight:
335.32
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.455

density

1.058 g/mL at 25 °C

SMILES string

CCCCCCCCCCCCOC(=O)C(C)(C)Br

InChI

1S/C16H31BrO2/c1-4-5-6-7-8-9-10-11-12-13-14-19-15(18)16(2,3)17/h4-14H2,1-3H3

InChI key

TUPZPNIRCIQNLF-UHFFFAOYSA-N

Application

Atom Transfer Radical Polymerization (ATRP) initiator for the creation of dodecyl functionalized telechelic polymers.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coessens, V.; Pintauer, T.; Matyjaszewski, K.
Progress in Polymer Science, 26, 337-337 (2001)
Atom transfer radical polymerization.
K Matyjaszewski et al.
Chemical reviews, 101(9), 2921-2990 (2001-12-26)
"Green" atom transfer radical polymerization: from process design to preparation of well-defined environmentally friendly polymeric materials.
Nicolay V Tsarevsky et al.
Chemical reviews, 107(6), 2270-2299 (2007-05-29)

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

Protocols

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service