Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723215

Sigma-Aldrich

10-Undecenyl 2-bromoisobutyrate

>95%

Synonym(s):

Allyl-type (long chain) initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27BrO2
CAS Number:
Molecular Weight:
319.28
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

>95%

refractive index

n20/D 1.450

density

1.080 g/mL at 25 °C

SMILES string

CC(C)(Br)C(=O)OCCCCCCCCCC=C

InChI

1S/C15H27BrO2/c1-4-5-6-7-8-9-10-11-12-13-18-14(17)15(2,3)16/h4H,1,5-13H2,2-3H3

InChI key

RHLOSBPISIVGMQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viktor Heinrichs et al.
PloS one, 13(4), e0195180-e0195180 (2018-04-07)
Living animal cells are strongly influenced by the mechanical properties of their environment. To model physiological conditions ultrasoft cell culture substrates, in some instances with elasticity (Young's modulus) of only 1 kPa, are mandatory. Due to their long shelf life
Coessens, V.; Pintauer, T.; Matyjaszewski, K.
Progress in Polymer Science, 26, 337-337 (2001)
Atom transfer radical polymerization.
K Matyjaszewski et al.
Chemical reviews, 101(9), 2921-2990 (2001-12-26)
"Green" atom transfer radical polymerization: from process design to preparation of well-defined environmentally friendly polymeric materials.
Nicolay V Tsarevsky et al.
Chemical reviews, 107(6), 2270-2299 (2007-05-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service