Skip to Content
MilliporeSigma
All Photos(3)

Documents

723185

Sigma-Aldrich

1,1,1-Tris(2-bromoisobutyryloxymethyl)ethane

97%

Synonym(s):

3f-BiB, Trifunctional initiator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27Br3O6
CAS Number:
Molecular Weight:
567.10
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

54-60 °C

SMILES string

CC(C)(Br)C(=O)OCC(C)(COC(=O)C(C)(C)Br)COC(=O)C(C)(C)Br

InChI

1S/C17H27Br3O6/c1-14(2,18)11(21)24-8-17(7,9-25-12(22)15(3,4)19)10-26-13(23)16(5,6)20/h8-10H2,1-7H3

InChI key

VLRDGSDXAISNJF-UHFFFAOYSA-N

Application

Atom Transfer Radical Polymerization (ATRP) initiator for the creation of trifunctional polymers.
Polymerization will occur at three sites creating a three-arm star polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Coessens, V.; Pintauer, T.; Matyjaszewski, K.
Progress in Polymer Science, 36, 337-337 (2001)
Atom transfer radical polymerization.
K Matyjaszewski et al.
Chemical reviews, 101(9), 2921-2990 (2001-12-26)
"Green" atom transfer radical polymerization: from process design to preparation of well-defined environmentally friendly polymeric materials.
Nicolay V Tsarevsky et al.
Chemical reviews, 107(6), 2270-2299 (2007-05-29)

Articles

ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.

Protocols

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service