Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

723061

Sigma-Aldrich

12-Mercaptododecanoic acid NHS ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27NO4S
Molecular Weight:
329.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

storage temp.

2-8°C

SMILES string

SCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O

InChI

1S/C16H27NO4S/c18-14-11-12-15(19)17(14)21-16(20)10-8-6-4-2-1-3-5-7-9-13-22/h22H,1-13H2

InChI key

WDHWRJMCVWOMCM-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service