Skip to Content
MilliporeSigma
All Photos(1)

Documents

722537

Sigma-Aldrich

4-Hydroxypiperidine-3,3,4,5,5-d5

98 atom % D, 98% (CP)

Synonym(s):

4-Piperidinol-3,3,4,5,5-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5H6NO
CAS Number:
Molecular Weight:
106.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

bp

108-114 °C/10 mmHg (lit.)

mass shift

M+5

SMILES string

[2H]C1([2H])CNCC([2H])([2H])C1([2H])O

InChI

1S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7H,1-4H2/i1D2,2D2,5D

InChI key

HDOWRFHMPULYOA-QJWYSIDNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service