Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

722030

Sigma-Aldrich

1-Boc-3-bromopyrrolidine

96%

Synonym(s):

tert-Butyl 3-bromopyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$149.00

$149.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$149.00

About This Item

Empirical Formula (Hill Notation):
C9H16BrNO2
CAS Number:
Molecular Weight:
250.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$149.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

96%

form

solid

mp

47-52 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCC(Br)C1

InChI

1S/C9H16BrNO2/c1-9(2,3)13-8(12)11-5-4-7(10)6-11/h7H,4-6H2,1-3H3

InChI key

QJTKPXFJOXKUEY-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service