Skip to Content
MilliporeSigma
All Photos(1)

Documents

721255

Sigma-Aldrich

2-(2-Methoxyethoxy)ethyl 2-(chlorosulfonyl)benzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16ClO6S
CAS Number:
Molecular Weight:
323.77
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.312

density

1.525 g/mL at 25 °C

SMILES string

COCCOCCOC(=O)c1ccccc1S(Cl)(=O)=O

InChI

1S/C12H15ClO6S/c1-17-6-7-18-8-9-19-12(14)10-4-2-3-5-11(10)20(13,15)16/h2-5H,6-9H2,1H3

InChI key

TVQUWKKLSJIKOI-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service