Skip to Content
MilliporeSigma
All Photos(1)

Documents

721190

Sigma-Aldrich

1,3-Bis(tributylstannyl)benzene

Synonym(s):

1,3-Bis(tributylstannanyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H58Sn2
CAS Number:
Molecular Weight:
656.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.520

density

1.148 g/mL at 25 °C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cccc(c1)[Sn](CCCC)(CCCC)CCCC

InChI

1S/C6H4.6C4H9.2Sn/c1-2-4-6-5-3-1;6*1-3-4-2;;/h1-3,6H;6*1,3-4H2,2H3;;

InChI key

ASPLJNIVVPATHK-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of covalent analogs of DNA base pairs and triplets from 9-benzyl-6-chloropurine via a Stille cross-coupling reaction

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service