Skip to Content
MilliporeSigma
All Photos(1)

Documents

720976

Sigma-Aldrich

Bis(11-cyanoundecyl) disulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H44N2S2
Molecular Weight:
424.75
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

30-34 °C

storage temp.

2-8°C

SMILES string

N#CCCCCCCCCCCCSSCCCCCCCCCCCC#N

InChI

1S/C24H44N2S2/c25-21-17-13-9-5-1-3-7-11-15-19-23-27-28-24-20-16-12-8-4-2-6-10-14-18-22-26/h1-20,23-24H2

InChI key

ROSJSPKGTQYEFY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service