Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720887

Sigma-Aldrich

Potassium dodecafluoro-closo-dodecaborate

97%

Synonym(s):

Dipotassium dodecafluorododecaborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B12F12K2
CAS Number:
Molecular Weight:
435.91
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: boron

mp

550 °C (dec.)

SMILES string

[K+].[K+].FB1C[B-]2(F)B(F)[B-](F)(B(F)B(F)B(F)B(F)B(F)B1F)B2F

InChI

1S/CH2B11F11.2K/c13-2-1-11(22)9(20)12(23,10(11)21)8(19)7(18)6(17)5(16)4(15)3(2)14;;/h1H2;;/q-2;2*+1

InChI key

MJZMONZFZXGMAR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service