Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

720429

Sigma-Aldrich

1H-Pyrazol-3-ylboronic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BN2O2 · xH2O
Molecular Weight:
111.90 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

88-93 °C

storage temp.

2-8°C

SMILES string

O.OB(O)c1cc[nH]n1

InChI

1S/C3H5BN2O2.H2O/c7-4(8)3-1-2-5-6-3;/h1-2,7-8H,(H,5,6);1H2

InChI key

NXQCQMBEFQLICU-UHFFFAOYSA-N

General description

1H-Pyrazol-3-ylboronic acid hydrate can be used as a reactant in the Suzuki-Miyaura coupling reaction to synthesize pyrazole derivatives by reacting with different aryl halides over the palladium catalyst.[1]

Application

Reactant for:
  • Preparation of receiver-amplifier, self-immolative dendrimers as potential platform for self-triggered multiple-release prodrugs
  • Preparation of bifunctional water-soluble triazole-containing derivatives of ethyl(hydroxycamptothecin) via click chemistry and conjugation to monoclonal antibodies for anticancer therapy

  • Intermediate for synthesis of oligo(ethylene glycol)-terminated alkanethiol amides useful for biosensors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Chemical Series of 5-Lipoxygenase-Activating Protein Inhibitors for Treatment of Coronary Artery Disease
Lemurell M, et al.
Journal of medicinal chemistry, 62(9), 4325-4349 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service