Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720216

Sigma-Aldrich

2-Methyl-3-nitrophenylacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO4
CAS Number:
Molecular Weight:
195.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
97%

Assay

97%

form

powder

mp

132-136 °C

storage temp.

2-8°C

SMILES string

Cc1c(CC(O)=O)cccc1[N+]([O-])=O

InChI

1S/C9H9NO4/c1-6-7(5-9(11)12)3-2-4-8(6)10(13)14/h2-4H,5H2,1H3,(H,11,12)

InChI key

LWIOFILTAJJDLA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service