Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

720143

Sigma-Aldrich

Potassium hydroxide on alumina

Synonym(s):

KOH on alumina

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HKO
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

>300 °C

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Application

Potassium hydroxide on alumina (KOH/AL2O3) can be employed as a base catalyst:
  • For the dehydrogenation of cyclic hydrocarbons (cycloolefins, cycloparaffins, and alkyl aromatics).
  • To prepare pyridazine derivatives from 1,2-dicarbonyls, and hydrazine hydrate under microwave (MW) irradiation.
  • In the total synthesis of natural product (-)-varitriol and (-)-3′-epi-varitriol (an isomer of (-)-varitriol) as potent antitumor agents.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas phase base-catalyzed dehydrogenations of cyclic hydrocarbons over a KOH/Al2O3 catalyst
Neumann R and Sasson Y
J. Mol. Catal., 35(1), 131-136 (1986)
The synthesis of (-)-varitriol and (-)-3?-epi-varitriol via a Ramberg-Backlund route
McAllister GD, et al.
Tetrahedron, 63(49), 12123-12130 (2007)
Potassium hydroxide impregnated Alumina (KOH-Alumina) as a recyclable catalyst for the solvent-free multicomponent synthesis of highly functionalized substituted pyridazines and/or substituted pyridazin-3 (2H)-ones under microwave irradiation
Mecadon H and Myrboh B
ISRN Organic Chemistry, 2011(1), 131-136 (2011)
Mizuki Takemoto et al.
PloS one, 10(6), e0131094-e0131094 (2015-06-27)
Channelrhodopsin (ChR) is a light-gated cation channel that responds to blue light. Since ChR can be readily expressed in specific neurons to precisely control their activities by light, it has become a powerful tool in neuroscience. Although the recently solved
Ashok R Patel et al.
Journal of agricultural and food chemistry, 63(19), 4862-4869 (2015-05-02)
The aim of this study was to use a detailed rheological characterization to gain new insights into the gelation behavior of natural waxes. To make a comprehensive case, six natural waxes (differing in the relative proportion of chemical components: hydrocarbons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service